Phenolphthalein, powder, BAKER ANALYZED® ACS suitable as pH-indicator, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

BAKR2870-01EA 105 SGD
BAKR2870-01
Phenolphthalein, powder, BAKER ANALYZED® ACS suitable as pH-indicator, J.T.Baker®
Phenolphthalein
Formula: C₂₀H₁₄O₄
Melting Pt: 258…262 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use Passes Test
ACS - Clarity of Solution Passes Test
Visual Transition Interval - pH…8.0 (Colorless) Passes Test
Visual Transition Interval - pH…10.0 (Red) Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR