J.T.Baker® DUAL-TINT®, pH Indicator Papers
Supplier: Avantor
BAKR2873-01EA 0 SGD
BAKR2873-01
J.T.Baker® DUAL-TINT®, pH Indicator Papers
Test Papers pH Indicator Papers and Test Strips
DUAL-TINT® pH papers offer easy-to-read, precise pH indication in a rolled 17-foot strip wide in a refillable dispenser with colour scale.
Two indicator zones are on one strip, separated by a white center division to give you clear colour gradations and reliable pH values.
Ordering information: Refills are avaialble for the dispenser in packs of three rolls.
Order Now
SPECIFICATIONS
Environmentally Preferable