Potassium hydrogen sulphate 90-100, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Potassium bisulphate

BAKR2960-01EA 120 SGD
BAKR2960-01 BAKR2960-05
Potassium hydrogen sulphate 90-100, BAKER ANALYZED®, J.T.Baker®
Potassium hydrogen sulphate

Lot analysis on label.

Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8 II
Merck Index: 13,07695

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Acidity (as H₂SO₄) 35.0 - 37.0 %
Insoluble Matter and NH₄OH Precipitate ≤0.01 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Calcium and Magnesium (as Ca) (by EDTA titration) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR