Potassium permanganate 99,0, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Permanganic acid potassium salt

BAKR3228-07EA 2620 SGD
BAKR3228-07
Potassium permanganate 99,0, BAKER ANALYZED® ACS, J.T.Baker®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
MDL Number: MFCD00011364
CAS Number: 7722-64-7
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (KMnO₄) ≥99.0 %
Insoluble Matter ≤0.2 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO₄) ≤0.02 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR