Sodium pyruvate ≥99.0% (by non aqueous titration), white powder, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor

Synonyms: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt

BAKR3354-02EA 317 SGD
BAKR3354-02 BAKR3354-04
Sodium pyruvate ≥99.0% (by non aqueous titration), white powder, BAKER ANALYZED®, J.T.Baker®
Sodium pyruvate

Lot analysis on label.

Formula: CH₃COCOONa
Storage Temperature: Refrigerator
MDL Number: MFCD00002586
CAS Number: 113-24-6
EINECS: 204-024-4

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 1% max.
Assay (CH₃COCOONa) (by non-aqueous titration) ≥99.0 %
Free Pyruvic Acid (CH₃COCOOH) ≤1 %
Solubility Passes Test
Meets Reagent Specifications for testing USP/NF monographs.
Store cold. Keep Dry. Warm to room temperature before opening.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR