Silver sulphate 98,0, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Disilver sulphate

BAKR3443-04EA 1180 SGD
BAKR3443-04
Silver sulphate 98,0, powder, BAKER ANALYZED® ACS, J.T.Baker®
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085
Melting Pt: 655
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements = 98.0 %
For Laboratory,Research,or Manufacturing Use = 0.02 %
ACS - Assay (Ag₂SO₄) ≥98.0 %
ACS - Insoluble Matter and Silver Chloride ≤0.02 %
ACS - Nitrate (NO₃) ≤0.001 %
ACS - Substances Not Precipitated by HCl ≤0.03 %
ACS - Iron (Fe) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR