Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Acetic acid sodium salt trihydrate

Management of Change (MOC) category = HR

BAKR3461-01EA 129 SGD
BAKR3461-01 BAKR3461-05 BAKR3461-07 BAKR3461-08 BAKR3461-19 BAKR3461-06 BAKR3461-10
Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Sodium acetate trihydrate

Optimize biopharmaceutical fermentation and cell culture productivity and improve the efficiency of your pharmaceutical production and biopharmaceutical processes with our proven-effective vitamins and pharmaceutical process minerals.


  • Food GMP manufactured product

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.45 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements 99.0 - 101.0%
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements ≤0.035%
Food GMP Manufactured Product ≤2.5
Meets J.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications ≤0.05%
Meets B.P. Chemical Specifications 38.0 - 41.0%
Preserve in tight containers, store at room temperature. 7.5 - 9.2
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical ≤0.005%
Bulk Pharmaceutical Chemical 99.0 - 101.0%
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤ 0.035 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
USP - Identification Passes Test
USP - Insoluble Matter ≤ 0.05 %
USP - Loss on Drying 38.0 - 41.0 %
USP - pH of 5% Solution at 25°C 7.5 - 9.2
USP - Potassium (K) Passes Test
USP - Sulfate (SO₄) ≤ 0.005 %
EP/BP - Assay (CH₃COONa) 99.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - pH (1 in 20) 7.5 - 9.0
EP/BP - Reducing Substances Passes Test
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Sulfate (SO₄) ≤ 200 ppm
EP/BP - Calcium and Magnesium ≤ 50 ppm
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying 39.0 - 40.5 %
JP - Assay (CH₃COONa) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Chloride (Cl) ≤ 0.011 %
JP - Sulfate (SO₄) ≤ 0.017 %
JP - Heavy Metals (as Pb) ≤ 10 ppm
JP - Calcium and Magnesium Passes Test
JP - Arsenic (As) ≤ 2 ppm
JP - Substances Reducing Permanganate Passes Test
JP - Loss on Drying 39.0 - 40.5 %
FCC - Alkalinity ≤ 0.05 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Loss on Drying 38.0 - 41.0 %
FCC - Potassium Compounds Passes Test
No Class 1, 2, 3 or other solvent is likely to be present in the final
product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR