Sodium acetate trihydrate 99.0-101.0% USP, FCC, J.T.Baker®

Supplier: Avantor

Synonyms: Acetic acid sodium salt trihydrate

BAKR3462-01EA 218 SGD
BAKR3462-01 BAKR3462-05 BAKR3462-R BAKR3462-07 BAKR3462-09
Sodium acetate trihydrate 99.0-101.0% USP, FCC, J.T.Baker®
Sodium acetate trihydrate

Crystals.

Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120…123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Food GMP Manufactured Product 99.0 - 101.0 %
Meets F.C.C. Requirements Passes Test
GMP Manufactured Product = 0.035 %
Meets U.S.P Requirements = 10 ppm
Preserve in tight containers, store at room temperature. Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical = 0.05 %
Bulk Food Chemical 38.0 - 41.0 %
USP - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
USP - Calcium and Magnesium Passes Test
USP - Chloride (Cl) ≤0.035 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Insoluble Matter ≤0.05 %
USP - Loss on Drying 38.0 - 41.0 %
USP - pH 7.5 - 9.2
USP - Potassium Compounds Passes Test
USP - Sulfate (SO₄) ≤0.005 %
FCC - Assay (CH₃COONa) (dried basis) 99.0 - 101.0 %
FCC - Alkalinity ≤0.05 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying 38.0 - 41.0 %
FCC - Potassium Compounds Passes Test
No Class 1, 2, 3 or other solvent is likely to be present in the final
product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR