Sodium acetate, anhydrous ≥99% USP, FCC, J.T.Baker®

Supplier: Avantor

Synonyms: Acetic acid sodium salt

Management of Change (MOC) category = R

BAKR3473-X1EA 17110 SGD
BAKR3473-X1
Sodium acetate, anhydrous ≥99% USP, FCC, J.T.Baker®
Sodium acetate

Caution: For Manufacturing, processing or repackaging.

Formula: H₃CCOONa
Melting Pt: 324 °C
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
Merck Index: 12,08711

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
Kit Components Meet Requirements Passes Test
This kit contains one each of Sodium Acetate, Anhydrous 3473-B6 and
3473-SB.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR