Sodium hydrogen carbonate kit 99.0-100.5% (dried basis), powder USP, FCC

Supplier: Avantor

Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

BAKR3509-K2EA 4240 SGD
BAKR3509-K2
Sodium hydrogen carbonate kit 99.0-100.5% (dried basis), powder USP, FCC
Sodium hydrogen carbonate

Caution: Not for use in hemodialysis.

Formula: NaHCO₃
Melting Pt: 270 °C
Density: 2,20 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655

Order Now

Specification Test Results

Meets F.C.C. Requirements
Meets U.S.P Requirement
GMP Manufactured Product
Food GMP Manufactured Product
Bulk Food Chemical
Bulk Pharmaceutical Chemica
Kit Components Meet Requirements Passes Test
This kit contains one each of Sodium Bicarbonate 3509-B2 and 3509-SA.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR