Sodium carbonate, granules, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

BAKR3604-01EA 158 SGD
BAKR3604-01 BAKR3604-05 BAKR3604-07
Sodium carbonate, granules, BAKER ANALYZED® ACS, J.T.Baker®
Sodium carbonate
Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay (Na₂CO₃) (dried basis)(by acidimetry) ≥99.5 %
Insoluble Matter ≤0.01 %
Loss on Heating at 285°C ≤1.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Silica (SiO₂) ≤0.005 %
Sulfur Compounds (as SO₄) ≤0.003 %
Calcium (Ca) ≤0.03 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR