Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®

Supplier: Avantor
Warning

BAKR3605-01EA 184 SGD
BAKR3605-01 BAKR3605-06 BAKR3605-09 BAKR3605-R
Sodium carbonate, anhydrous 99.5-100.5% (by anhydrous basis), granular NF, FCC, J.T.Baker®
Sodium carbonate

pH adjusters are used to adjust the pH of formulations.


  • Food GMP manufactured product

Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product Passes Test
Food GMP Manufactured Product Passes Test
Meets F.C.C. Requirements ≤0.5%
Meets N.F. Requirements 99.5 - 100.5%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical ≤0.5%
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) ≤0.5 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Loss on Drying ≤0.5 %
FCC - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
FCC - Lead (Pb) ≤4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR