Sodium carbonate 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

Management of Change (MOC) category = R

BAKR3606-05EA 324 SGD
BAKR3606-05 BAKR3606-09 BAKR3606-R BAKR3606-07
Sodium carbonate 99.5-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Sodium carbonate

  • GMP manufactured product

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets N.F. Requirements Passes Test
Meets J.P. Chemical Specifications Passes Test
Meets B.P. Chemical Specifications ≤0.5%
Meets E.P. Chemical Specifications 99.5 - 100.5%
GMP Manufactured Product 99.5 - 100.5%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Water (H₂O) ≤ 0.5 %
NF - Assay (Na₂CO₃) (anhydrous basis) 99.5 - 100.5 %
EP/BP - Assay (Na₂CO₃) (dried basis) 99.5 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Identification D Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Alkali Hydroxides and Bicarbonates Passes Test
EP/BP - Chloride (Cl) ≤ 125 ppm
EP/BP - Sulfate (SO₄) ≤ 250 ppm
EP/BP - Iron (Fe) ≤ 50 ppm
EP/BP - Loss on Drying ≤ 1.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤ 0.071 %
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Arsenic (As) ≤ 3.1 ppm
JP - Loss on Drying ≤ 0.5 %
JP - Assay (Na₂CO₃) (dried basis) 99.0 - 101.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR