Sodium chloride ≥99.0% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

BAKR3624-20EA 668 SGD
BAKR3624-20 BAKR3624-01 BAKR3624-05 BAKR3624-19 BAKR3624-06 BAKR3624-07 BAKR3624-60
Sodium chloride ≥99.0% (by argentometric titration), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements ≥99.0%
For Laboratory,Research,or Manufacturing Use 5.0 - 9.0
Assay (NaCl) (by Ag titrn) ≥99.0 %
pH of 5% Solution at 25°C 5.0 - 9.0
Insoluble Matter ≤0.005 %
Iodide (I) ≤0.002 %
Bromide (Br) ≤0.01 %
Chlorate and Nitrate (as NO₃) ≤0.003 %
ACS - Phosphate (PO₄) ≤5 ppm
Sulfate (SO₄) ≤0.004 %
Barium (Ba) Passes Test
ACS - Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) ≤2 ppm
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.005 %
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR