Sodium chloride 99.0-100.5% (dry basis), granular USP, FCC

Supplier: Avantor
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

Management of Change (MOC) category = R

BAKR3628-X2EA 7990 SGD
BAKR3628-X2
Sodium chloride 99.0-100.5% (dry basis), granular USP, FCC
Sodium chloride

Certifications: Must be subjected to further processing during the preparation of injectable dosage forms. Meets USP and FCC requirements. GMP manufactured product.


Caution: Preserve in tightly sealed containers.

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2,16 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Food GMP Manufactured Product
Meets F.C.C. Requirements
GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
Kit Components Meet Requirements Passes Test
This kit contains four (4) each of Sodium Chloride 3628-B6 and 3628-SA.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR