Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

BAKR3764-01EA 312 SGD
BAKR3764-01
Sodium molybdate(VI) dihydrate 99.5-103.0%, BAKER ANALYZED® ACS, J.T.Baker®
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3,28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (Na₂MoO₄·2H₂O) 99.5 - 103.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 7.0 - 10.5
Chloride (Cl) ≤0.005 %
Ammonium (NH₄) ≤0.001 %
Sulfate (SO₄) ≤0.015 %
Trace Impurities - Iron (Fe) ≤0.001 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR