J.T.Baker®, Formalin Neutraliser
Supplier: Avantor
BAKR3934.5000EA 140 SGD
BAKR3934.5000
J.T.Baker®, Formalin Neutraliser
Stains Microscopy and staining reagents
Formalin (dissolved formaldehyde) is used in histopathology laboratories.
  • Histology grade powdered chemical
  • Contains sodium disulphite min. 80 %
  • Stable for two years when bottles are closed and stored at room temperature
Packaging: In a PP drum.
Order Now
SPECIFICATIONS
Environmentally Preferable