Sodium chloride ≥99.0%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

BAKR4058-01EA 74.1 SGD
BAKR4058-01 BAKR4058-02
Sodium chloride ≥99.0%, Ultrapure Bioreagent for liquid chromatography, for molecular biology, J.T.Baker®
Sodium chloride

ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (NaCl) ≥99.0 %
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
pH of 5% Solution at 25°C 5.5 - 7.5
Insoluble Matter ≤0.005 %
Bromide (Br) ≤0.01 %
Chlorate and Nitrate (as NO₃) ≤0.003 %
Iodide (I) ≤0.002 %
ACS - Sulfate (SO₄) ≤0.003 %
Barium (Ba) ≤0.001 %
Calcium, Magnesium, and R₂O₃ Precipitate ≤0.005 %
Potassium (K) ≤0.005 %
Nitrogen Compounds (as N) ≤5 ppm
Phosphate (PO₄) ≤5 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR