Agarose PFGE, Ultrapure Bioreagent, J.T.Baker®

Supplier: Avantor

BAKR4063-05EA 0 SGD
BAKR4063-05
Agarose PFGE, Ultrapure Bioreagent, J.T.Baker®
Agarose

For rapid resolution of DNA greater than 1000 bp

Density: 0,90 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Loss on Drying at 70°C (in vacuo) ≤7.0 %
Sulfate (SO₄) ≤0.12 %
Electroendosmosis (EEO)(-m^r) ≤0.12 %
Gel Strength, g/cm² (MCl, 1.0%) ≥2000
Gelling Temperature of 1.5% w/w Solution 34.5 - 37.5 °C
Melting Temperature of 1.5% w/w Solution 86.5 - 89.5 °C
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR