Guanidinium thiocyanate ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

BAKR4083-01EA 0 SGD
BAKR4083-01
Guanidinium thiocyanate ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118 °C
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013027
CAS Number: 593-84-0
EINECS: 209-812-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NH₂C(:NH)NH₂·HSCN) (dried basis) ≥99.0 %
Identification Passes Test
Loss on Drying at 105°C ≤0.5 %
pH of 5% Solution at 25°C 4.0 - 7.0
Absorbance of a 1.7M Solution - 340 nm ≤0.030
Absorbance of a 1.7M Solution - 300 nm ≤0.050
Absorbance of a 1.7M Solution - 280 nm ≤0.300
DNase Activity (None Detected) Passes Test
Protease Activity (None Detected) Passes Test
RNase Activity (None Detected) Passes Test
Trace Impurities - Arsenic (As) ≤1 ppm
Trace Impurities - Copper (Cu) ≤1 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Lead (Pb) ≤2.000 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR