Sodium dodecyl sulphate (SDS) 99, ultrapure_bioreagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

BAKR4095-07EA 4730 SGD
BAKR4095-07 BAKR4095-02 BAKR4095-04 BAKR4095-05
Sodium dodecyl sulphate (SDS) 99, ultrapure_bioreagent, J.T.Baker®
Sodium dodecyl sulphate (SDS)

For electrophoresis and protein solubilization.


  • Lot analysis on label

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 0.40 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2926
ADR: 4.1, 6.1 II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥99%
Appearance Passes Test
Assay (as C₁₂H₂₅OSO₃Na) ≥99 %
Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path) ≤0.1
pH of 1% Solution at 25°C 5.0 - 8.0
Chloride (Cl) ≤0.03 %
Nitrogen (N) ≤5 ppm
Phosphate (PO₄) ≤1 ppm
Heavy Metals (as Pb) ≤2 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR