L(+)-Tartaric acid, granular NF, Multi-Compendial, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid

BAKR4105-05EA 321 SGD
BAKR4105-05
L(+)-Tartaric acid, granular NF, Multi-Compendial, J.T.Baker®
L(+)-Tartaric acid
Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 399
Melting Pt: 168…170 °C
Density: 1.759 g/cm³ (20 °C)
Flash Pt: 150 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00064207
CAS Number: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
Meets J.P. Chemical Specifications
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+) 12.0 - 13.0 Degree
NF - Loss on Drying ≤0.5 %
NF - Residue on Ignition ≤0.1 %
NF - Oxalate (C₂O₄) Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Assay (HOCO(CHOH)₂COOH) (dried basis) 99.7 - 100.5 %
NF - Endotoxin Concentration (EU/g) ≤10
EP/BP - Assay (HOCO(CHOH)₂COOH) (dried basis) 99.5 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation [Å]²º^D (+) 12.0 - 12.8 Degree
EP/BP - Oxalic Acid ≤360 ppm
EP/BP - Chloride (Cl) ≤100 ppm
EP/BP - Sulfate (SO₄) ≤150 ppm
EP/BP - Calcium (Ca) ≤200 ppm
EP/BP - Loss on Drying at 105°C ≤0.2 %
EP/BP - Ash (sulfated) ≤0.1 %
JP - Assay (HOCO(CHOH)₂COOH) (dried basis) 99.7 - 101.0 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Sulfate (SO₄) ≤0.048 %
JP - Oxalate (C₂O₄) Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Calcium (Ca) Passes Test
JP - Arsenic (As) ≤1 ppm
JP - Loss on Drying ≤0.5 %
JP - Residue on Ignition ≤0.05 %
No Class 1,2,3 or other solvents are used or produced in the = 0.05 %
manufacturing or purification of the product.
The USP Organic Volatile Impurities <467> requirements for this product is met by the implementation of Residual Solvents <467> as per the USP Revision Bulletin of June 15, 2007.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR