Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: Avantor


Management of Change (MOC) category = NI

BAKR4218-03EA 125 SGD
BAKR4218-03 BAKR4218-02 BAKR4218-88_U BAKR4218-88 BAKR4218-P04
Water, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 0.0010
UV Absorbance on Gradient Elution - Largest Peak at 254nm (au) ≤0.001
UV Absorbance on Gradient Elution - Largest Peak at 205nm (au), for
Information Only = 0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤0.2 ppb
Filtration Test Passes Test
pH, For Information Only = 2.0 ppm
Residue after Evaporation ≤2.0 ppm
Filtered through a 0.2 micron filter.
Appearance - A Clear, Colourless, Odourless Liquid
Appearance - A clear, colorless, odorless liquid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR