Water, Ultrapure Bioreagent for molecular biology, sterile, J.T.Baker®

Supplier: Avantor

BAKR4221-02EA 108 SGD
BAKR4221-02
Water, Ultrapure Bioreagent for molecular biology, sterile, J.T.Baker®
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Sterilized with diethylpyrocarbonate (DEPC) for nucleic acid applications


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR