J.T.Baker® Plate, Dilution Strips SW
Supplier: Avantor
BAKR43001-0031EA 293 SGD
BAKR43001-0031
J.T.Baker® Plate, Dilution Strips SW
Microplates Deep Well Microplates
Suitable for use on Dynex DSX and DS2 instruments. Deep well reservoir with eight wells.
  • Strip of 8 wells
  • Made of PP