J.T.Baker® Plate, SRW Magnetic
Supplier: Avantor
BAKR43001-0505EA 599 SGD
BAKR43001-0505 BAKR43001-0504 BAKR43001-0506 BAKR43001-0514 BAKR43001-0515 BAKR43001-0516
J.T.Baker® Plate, SRW Magnetic
Microplates HTS Microplates
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Apex, Flex 96 and Presto.
  • Standard SBS footprint
  • Cylindrical wells enable good sample mixing
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code assists registration and identification of individual samples
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU) ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU) ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU) ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU