Zinc sulphate heptahydrate ≥99.0%, granular USP, ACS, J.T.Baker®

Supplier: Avantor
Danger

BAKR4372-01EA 325 SGD
BAKR4372-01
Zinc sulphate heptahydrate ≥99.0%, granular USP, ACS, J.T.Baker®
Zinc sulphate heptahydrate
Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100
MDL Number: MFCD00149894
CAS Number: 7446-20-0
UN: 3077
ADR: 9 III
Merck Index: 12,10293

Order Now

Specification Test Results

GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.0 - 108.7 %
USP - Assay (ZnSO₄·7H₂O) 99.0 - 108.7 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Acidity Passes Test
USP - Alkalies and Alkaline Earths ≤0.5 %
USP - Arsenic (As) ≤3 ppm
USP - Lead (Pb) (USP) ≤0.002 %
ACS - Ammonium (NH₄) ≤0.001 %
ACS - Assay (ZnSO₄·7H₂O) 99.0 - 103.0 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Lead (Pb) ≤0.003 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Nitrate (NO₃) ≤0.002 %
ACS - pH of 5% Solution at 25°C 4.4 - 6.0
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Manganese (Mn) ≤2.000 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR