J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
Supplier: Avantor
BAKR4467-02EA 0 SGD
BAKR4467-02 BAKR4480-04 BAKR4467-04
J.T.Baker® BAKER-flex®, TLC Sheets, Precoated
TLC Plates
Baker-flex® pre-coated flexible polyester sheets have a hard, abrasion resistant coating and solvent resistant substrate for rapid, clean separations. The sheets can be written on and cut to size.
Order Now
SPECIFICATIONS
Environmentally Preferable