J.T.Baker®, Robotic Tips Type ST
Supplier: Avantor
BAKR49000-0000EA 3370 SGD
BAKR49000-0000 BAKR49000-0006 BAKR49001-0000 BAKR49001-0106 BAKR49000-1106 BAKR49001-1106 BAKR49001-3106 BAKR49000-2000
J.T.Baker®, Robotic Tips Type ST
Pipette Tips
Suitable for use on Euroimmun Analyzer. Available in conductive version, with or without filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • Conductive tips for liquid level sensing
  • Ideal for dosing even small volumes with high precision and without contamination
J.T.Baker® robotic tips type ST are suitable for use on Euroimmun Analyzer and other robotic instruments using the same pipetting head.
The validation of the tips on the instrument for specific applications isMore Product Information