J.T.Baker®, Robotic Tips Type QG
Supplier: Avantor
BAKR49018-0106EA 646 SGD
BAKR49018-0106
J.T.Baker®, Robotic Tips Type QG
Pipette Tips
Suitable for use on third party robotic instruments QIAgility. Available in conductive version, with filter, non-sterile, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • State of the art quality control
  • Free of DNA, DNase, RNase and PCR inhibitors
  • Ideal for high-throughput analysis and DNA purification
  • Packed in blister
The validation of the tips on the instrument for specific applications is within the responsibility of the user.