J.T.Baker®, Robotic Tips Type MP
Supplier: Avantor
BAKR49209-0105EA 1150 SGD
BAKR49209-0105
J.T.Baker®, Robotic Tips Type MP
Pipette Tips
Suitable for use with Roche MagNA Pure instruments. Available in conductive version, with filter, and in CE-IVD marked version for clinical diagnostic applications.
  • Manufactured under cleanroom conditions (cleanroom Class 8)
  • State-of-the-art quality control
  • Barcoded trays for automated identification
  • Free of DNA, DNase, RNase and PCR inhibitors
  • Ideal for high throughput analysis
  • Packed in blisters
The validation of the tips on the instrument for specific applications is within the responsibility of the user.