J.T.Baker® Robotic Tips Type TE MCA
Supplier: Avantor
BAKR49306-1109EA 1440 SGD
BAKR49306-1109 BAKR49332-0009 BAKR49306-1008 BAKR49306-0009 BAKR49305-0009 BAKR49306-0008 BAKR49304-1109 BAKR49304-1008 BAKR49304-0009 BAKR49304-0008 BAKR49302-1109 BAKR49302-1008 BAKR49302-0008 BAKR49302-0009
J.T.Baker® Robotic Tips Type TE MCA
Pipette Tips
J.T.Baker Robotic tips type TE MCA are compatible with the Tecan Freedom EVO and the Tecan Fluent liquid handler using the MCA 96 head. Tips are manufactured under automated processes to reduce human contact and ensure precise reproducibility and exceptional quality.
  • Tested and certified free of human DNA, DNase and RNase, BSE/TSE, PCR inhibitors, ATP, and heavy metals
  • Manufactured in an ISO 9001 & ISO 13485 registered facility under cleanroom (class 8) conditions
  • Tips are made with virgin polypropylene ensuring your lab automation is free from resin
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)