n-Hexane, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Supplier: Avantor

Synonyms: Hexyl hydride, Hexanen, Hexane

BAKR5274.2500EA 99.2 SGD
BAKR5274.2500
n-Hexane, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Flash Pt: -22
MDL Number: MFCD00009520
CAS Number: 110-54-3
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 95.0 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR