Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®

Supplier: Avantor

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

BAKR5279.2500EA 72.8 SGD
BAKR5279.2500
Methanol, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Methanol
Formula: CH₃OH
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99.9 %
Acidity max. 0.0005 meq/g
Evaporation Residue max. 5 ppm
GC sensitive impurities (lindane GC-ECD) max. 5 ng/l
Water (KF) max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR