Cerium(IV) sulphate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: Avantor
Danger

BAKR5626-02EA 156 SGD
BAKR5626-02
Cerium(IV) sulphate 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Cerium (IV) sulphate

Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.

Formula: Ce(SO₄)₂
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8,II
Merck Index: 12,02036

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.095 - 0.105
Normality 0.095 - 0.105
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR