Potassium permanganate 0.0995-0.1005 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®

Supplier: Avantor

Synonyms: Permanganic acid potassium salt

BAKR5651-02EA 65 SGD
BAKR5651-02 BAKR5651-03
Potassium permanganate 0.0995-0.1005 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Potassium permanganate

Standardised at 25 °C against chemicals whose certification is traceable to NIST. Lot standardisation value, accurate to ±1 part per 1000, provided on label.

Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9 III
Merck Index: 12,07824

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005
Chloride and Chlorate (as Cl) ≤10 ppm
Sulfate (SO₄) ≤20 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.
SRM No…


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR