Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®

Supplier: Avantor
Danger

BAKR5907-01EA 73.1 SGD
BAKR5907-01
Barium chloride T.S., BAKER ANALYZED® PharmaTest™, J.T.Baker®
Barium chloride
Formula: BaCl₂
Density: 1,00 g/cm³
MDL Number: MFCD00003445
CAS Number: 10361-37-2
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (BaCl₂·2H₂O), w/v 11.6 - 12.4 %
Meets test solution (T.S.) specifications for testing USP/NF monographs.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR