J.T.Baker® BAKER INSTRA-ANALYZED®, Aluminium AA Standard
Supplier: Avantor
BAKR6440-04EA 149 SGD
BAKR6440-04
J.T.Baker® BAKER INSTRA-ANALYZED®, Aluminium AA Standard
Standards Cationic Standards Aluminium Standards
The standard is prepared by the careful dilution of the metal or its salt, usually 99,99% pure as verified by arc emission spectrometry.
  • Standardised with NIST standard chemicals
  • Maximum shelf life and stability
The final concentration of each lot is determined by titrimetry or gravimetry.
Order Now
SPECIFICATIONS
Environmentally Preferable