J.T.Baker® BAKERBOND spe™ Cyano (CN), SPE Columns
Supplier: Avantor
BAKR7021-03EA 0 SGD
BAKR7021-03
J.T.Baker® BAKERBOND spe™ Cyano (CN), SPE Columns
Chromatography Columns
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Non ionic, non polar to polar analytes.
These SPE columns can be used in reversed phase and normal phase mode.