J.T.Baker®, Silica Gels for Preparative Chromatography, Low Pressure
Supplier: Avantor
BAKR7024-01EA 654 SGD
BAKR7024-01
J.T.Baker®, Silica Gels for Preparative Chromatography, Low Pressure
Chromatography Sorbents HPLC Sorbents
These are used in low pressure preparative chromatography. The mesh specifications ensure there is no excess of fines in the product which can affect flow rates and separation performance.
  • High surface area improves productivity
  • Narrow pore size distribution sharpens peaks and improves resolution