J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Supplier: Avantor
BAKR7061-00EA 101 SGD
BAKR7061-00
J.T.Baker® BAKERBOND®, Florisil Bulk Packing Material
Chromatography Sorbents SPE Sorbents
SPE sorbent for clean-up columns. Suitable for the determination of Hydrocarbon Oil Index according ISO 9377-2 and NEN 5733.