J.T.Baker® BAKERBOND spe™, Filtration Columns with Frits
Supplier: Avantor
BAKR7121-06EA 445 SGD
BAKR7121-06
J.T.Baker® BAKERBOND spe™, Filtration Columns with Frits
Chromatography Columns
These empty filtration SPE columns are packed with dual 20 µm frits. They are designed for use with spe-12G and spe-24G glass column processors.
Order Now
SPECIFICATIONS
Environmentally Preferable