J.T.Baker® BAKERBOND spe™ Carboxylic Acid, SPE Columns
Supplier: Avantor
BAKR7211-03EA 871 SGD
BAKR7211-03
J.T.Baker® BAKERBOND spe™ Carboxylic Acid, SPE Columns
Chromatography Columns
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Ionic, basic analytes.
These SPE columns can be used in weak cation exchange mode.