J.T.Baker® BAKERBOND spe™ Florisil, SPE Columns
Supplier: Avantor
BAKR7420-07EA 0 SGD
BAKR7420-07
J.T.Baker® BAKERBOND spe™ Florisil, SPE Columns
Chromatography Columns
BAKERBOND spe™ columns are disposable PP or glass columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum or positive pressure
General applications: Low to moderately polar analytes from non aqueous solutions.
These SPE columns can be used in adsorption mode.
Packaging: Certain columns are available in a foil-lined fibre drum called the Jumbo Pack. The columns are loose packed in the foilMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable