J.T.Baker® BAKERBOND spe™ Carbon-Amino, SPE Columns for Special Applications
Supplier: Avantor
BAKR7450-07EA 761 SGD
BAKR7450-07
J.T.Baker® BAKERBOND spe™ Carbon-Amino, SPE Columns for Special Applications
Chromatography Columns
BAKERBOND spe™ Amino-Carbon is a double-phase solid extraction column packed with spherical activated carbon (lower layer) and aminopropyl modified silica phase (upper layer) of polar, weak anion properties. Designed for pesticide residue analysis in agricultural product.