Chlorobenzene, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

BAKR8023.2500EA 112 SGD
BAKR8023.2500
Chlorobenzene, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1,11 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99 %
Boiling Range max. 1.0 °C
Density (g/ml) at 25°C 1.101 - 1.104
Recorded Boiling Point 132 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR