J.T.Baker® BAKERBOND Speedisk®, Extraction Stations
Supplier: Avantor
BAKR8095-06EA 11530 SGD
BAKR8095-06
J.T.Baker® BAKERBOND Speedisk®, Extraction Stations
Manifolds
The BAKERBOND Speedisk® expanded extraction station can be used in reservoir, inverted, or remote sample feed modes. It includes a six-port vacuum manifold and the accessories needed to support the extraction of analyte by BAKERBOND Speedisk® laminar extraction discs.
The manifold has a rectangular footprint and inter-port spacing to accommodate six, side-by-side, 1 L sample reservoirs. Each vacuum port has an individual open/close valve.
Delivery information: Including extraction station, two remote sample adapters, two collection chambers and two vials, six reservoirs of 185 ml.
Order Now
SPECIFICATIONS
Environmentally Preferable