Ethanol absolute, BAKER ANALYZED® HPLC for HPLC, analytical reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Ethyl alcohol

BAKR8462.2500EA 79.3 SGD
BAKR8462.2500
Ethanol absolute, BAKER ANALYZED® HPLC for HPLC, analytical reagent, J.T.Baker®
Ethanol
Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78.3
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay min. 99.5 %
Density (g/ml) at 20°C 0.789 - 0.790
Free Acid (as CH3COOH) max. 0.001 %
Residue after Evaporation max. 0.001 %
Water (H2O) max. 0.2 %
Ultraviolet Absorbance (1.00-cm path vs water):
at 210 nm max. 0.7
at 240 nm max. 0.1
at 260 nm max. 0.01

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR