Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®

Supplier: Avantor
Danger

BAKR9018-03EA 336 SGD
BAKR9018-03
Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤ 0.010
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤ 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm ≤ 0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤ 190
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤ 0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤ 1.0 ppb
Appearance Passes Test
Assay (CH₃CN) (by GC, corrected for water) ≥ 99.8 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 1.0 ppm
Titrable Acid (µeq/g) ≤ 0.8
Titrable Base (µeq/g) ≤ 0.6
Water (by KF, coulometric) ≤ 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR