Benzyl alcohol 98.0-100.5% NF, J.T.Baker®

Supplier: Avantor
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

BAKR9040-03EA 480 SGD
BAKR9040-03
Benzyl alcohol 98.0-100.5% NF, J.T.Baker®
Benzyl alcohol

Caution: This product is not intended for use in preparing parenteral dosage forms.

Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.4…–15.4 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 100.40 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 98.0 - 100.5 %
NF - Assay 98.0 - 100.5 %
NF - Clarity of Solution Passes Test
NF - Color of Solution Passes Test
NF - Identification Passes Test
NF - Peroxide Value ≤5
NF - Refractive Index, n²ºD 1.538 - 1.541
NF - Benzaldehyde and Other Related Substances - Benzaldehyde ≤0.15 %
NF - Related Substances - Cyclohexylmethanol ≤0.10 %
of Benzyl Alcohol ≤0.04 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤0.04 %
NF - Benzaldehyde and Other Related Substances - Total Other Peaks ≤0.3 %
Greater Than RT of Benzyl Alcohol ≤0.3 %
NF - Acidity Passes Test
NF - Residue on Evaporation ≤0.05 %
NF - Benzene ≤2 ppm
This product is not intended for use in preparing parenteral dosage
forms.
The Class 1 solvent Benzene is likely to be present below 2 ppm. The
actual analysis of Benzene is reported for each lot.
The Class 2 solvent Toluene is likely to be present and is process
controlled to <30 ppm.
No other Class 1, 2 or 3 residual solvents are likely to be present.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR